Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177615.3 | NP_808283.2 | 4 | 715 | ||
GenBank mRNA | AK052641.1 | - | 654 | |||
BC089587.1 | - | 4 | 652 | 6757541 |