Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Engase | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172573.2 | NP_766161.1 | 4 | 471 | ||
GenBank mRNA | AK048667.1 | - | 4 | 473 | ||
AK084254.1 | - | 4 | 604 | |||
AK167142.1 | - | 4 | 443 | |||
BC120923.1 | - | 4 | 469 | |||
BC120924.1 | - | 4 | 473 | 40097903 |