Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081654.2 | NP_001075123.1 | 3 | 361 | ||
NM_001286498.1 | NP_001273427.1 | 3 | 561 | |||
NM_011573.3 | NP_035703.2 | 3 | 816 | |||
GenBank mRNA | AK003113.1 | - | 3 | 324 | ||
AK009326.1 | - | 3 | 667 | |||
AK159046.1 | - | 2 | 1181 | |||
AK219517.1 | - | 1 | 94 | |||
BB860146.1 | - | 3 | 361 | |||
BC002248.1 | - | 3 | 325 | 3493141 | ||
BC006608.1 | - | 3 | 299 | 3157453 | ||
CX220909.1 | - | 3 | 329 | |||
DV661702.1 | - | 1 | 136 |