Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148926.2 | NP_683728.1 | 4 | 487 | ||
GenBank mRNA | AK082915.1 | - | 3 | 483 | ||
AK148316.1 | - | 516 | ||||
AK160675.1 | - | 5 | 410 | |||
BC024483.1 | - | 3 | 148 | |||
BC027161.1 | - | 3 | 285 | |||
BC034396.1 | - | 4 | 407 | 3707476 | ||
BC083124.1 | - | 4 | 487 | 6334368 | ||
BC100559.1 | - | 5 | 404 | 3419074 | ||
X80437.1 | - | 5 | 383 |