Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172582.3 | NP_766170.2 | 11, 12 | 1488 | ||
GenBank mRNA | AK077863.1 | - | 11, 12 | 1412 | ||
AK131889.1 | - | 11, 12 | 1450 | |||
AK167380.1 | - | 11, 12 | 1416 | |||
AK170311.1 | - | 10, 11 | 1953 | |||
AK191776.1 | - | 2, 3 | 258 | |||
BC024135.1 | - | 11, 12 | 1389 | |||
BC051055.1 | - | 11, 12 | 1404 | |||
BC057914.1 | - | 8, 9 | 1152 | |||
BC088736.1 | - | 7, 8 | 2737 | |||
BC106183.1 | - | 11, 12 | 1472 | |||
BC132168.1 | - | 11, 12 | 1488 | 40130445 | ||
BC132170.1 | - | 11, 12 | 1488 | 40130447 |