Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146198.1 | NP_001139670.1 | 2 | 451 | ||
NM_009385.3 | NP_033411.3 | 3 | 1018 | |||
GenBank mRNA | BC057607.1 | - | 3 | 1018 | 6416507 | |
BC080868.1 | - | 2 | 498 | 30363051 |