Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145465.2 | NP_663440.1 | 9 | 1376 | ||
GenBank mRNA | AK154596.1 | - | 9 | 1481 | ||
AK154959.1 | - | 9 | 1377 | |||
AY188357.1 | - | 9 | 1386 | |||
BC004650.1 | - | 9 | 1208 | 3482980 |