Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163518.1 | NP_001156990.1 | 1 | 960 | ||
NM_146059.2 | NP_666171.2 | 2 | 847 | |||
GenBank mRNA | AK137409.1 | - | 1 | 660 | ||
AK171003.1 | - | 1 | 631 | |||
BC024536.1 | - | 1 | 568 | 4988004 | ||
BC047213.1 | - | 1 | 961 | 6395374 |