Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Treml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033922.2 | NP_001029094.1 | 2 | 327 | ||
NM_001163795.1 | NP_001157267.1 | 2 | 339 | |||
NM_001163796.1 | NP_001157268.1 | 2 | 339 | |||
NM_172623.2 | NP_766211.2 | 2 | 339 | |||
GenBank mRNA | AK077228.1 | - | 2 | 339 | ||
AK156085.1 | - | 2 | 342 | |||
AY465530.1 | - | 2 | 297 | |||
AY465531.1 | - | 2 | 297 | |||
AY465532.1 | - | 2 | 297 | |||
AY557629.1 | - | 2 | 275 | |||
BC117091.1 | - | 2 | 314 | 40126410 | ||
BC137666.1 | - | 2 | 314 | 8860685 | ||
DQ087185.1 | - | 2 | 261 | |||
DQ087186.1 | - | 2 | 261 | |||
DQ186654.1 | - | 2 | 281 | |||
DQ186655.1 | - | 2 | 293 |