Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204914.1 | NP_001191843.1 | 6, 7 | 717 | ||
NM_144865.2 | NP_659114.2 | 6, 7 | 723 | |||
GenBank mRNA | AK158694.1 | - | 1, 2 | 297 | ||
AY562230.1 | - | 6, 7 | 556 | |||
BC020184.1 | - | 6, 7 | 654 | 4505679 | ||
EU555417.1 | - | 6, 7 | 550 |