Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204914.1 | NP_001191843.1 | 7 | 823 | ||
NM_144865.2 | NP_659114.2 | 7 | 829 | |||
GenBank mRNA | AK147333.1 | - | 1 | 5619 | ||
AK158694.1 | - | 2 | 403 | |||
AY562230.1 | - | 7 | 662 | |||
BC020184.1 | - | 7 | 760 | 4505679 | ||
EU555417.1 | - | 7 | 656 |