Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271443.1 | NP_001258372.1 | 6 | 502 | ||
NM_001271444.1 | NP_001258373.1 | 6 | 742 | |||
NM_144870.5 | NP_659119.2 | 6 | 745 | |||
GenBank mRNA | AK135069.1 | - | 6 | 503 | ||
AK149674.1 | - | 6 | 703 | |||
AK153182.1 | - | 6 | 689 | |||
AK153900.1 | - | 6 | 479 | |||
AK164901.1 | - | 6 | 479 | |||
AK167907.1 | - | 6 | 480 | |||
BC021616.1 | - | 6 | 463 | 5132866 | ||
BC086766.1 | - | 6 | 456 | 6820786 | ||
BP775598.1 | - | 2 | 334 |