Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175013.2 | NP_778178.3 | 1, 2 | 316 | ||
GenBank mRNA | AK035507.1 | - | 1, 2 | 316 | ||
AK076484.1 | - | 1, 2 | 162 | |||
BC115937.1 | - | 1, 2 | 126 | 40089650 |