Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139236.3 | NP_631982.2 | 4, 5 | 685 | ||
GenBank mRNA | AF361077.1 | - | 4, 5 | 634 | ||
AF361078.1 | - | 4, 5 | 634 | |||
AK155818.1 | - | 4, 5 | 624 | |||
AK166559.1 | - | 4, 5 | 611 | |||
AK166650.1 | - | 4, 5 | 611 | |||
AK171163.1 | - | 4, 5 | 622 | |||
AK171421.1 | - | 4, 5 | 612 | |||
BC059820.1 | - | 4, 5 | 603 | 6827596 | ||
BC060189.1 | - | 4, 5 | 613 |