Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172723.4 | NP_766311.2 | 10, 11 | 1205 | ||
GenBank mRNA | AK076754.1 | - | 9, 10 | 1191 | ||
AK076768.1 | - | 10, 11 | 962 | |||
BC056934.1 | - | 9, 10 | 931 | |||
BC079634.1 | - | 9, 10 | 999 | |||
BC144874.1 | - | 9, 10 | 947 |