Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172279.1 | NP_758483.1 | 7 | 517 | ||
GenBank mRNA | AK122565.2 | - | 3 | 163 | ||
AK146784.1 | - | 8 | 852 | |||
AY151083.1 | - | 7 | 517 |