Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011874.2 | NP_036004.2 | 6, 7 | 741 | ||
GenBank mRNA | AK050677.1 | - | 6, 7 | 719 | ||
AK077507.1 | - | 6, 7 | 704 | |||
AK150989.1 | - | 6, 7 | 696 | |||
AK153991.1 | - | 6, 7 | 724 | |||
AK160644.1 | - | 6, 7 | 703 | |||
AK160718.1 | - | 6, 7 | 695 | |||
AK167041.1 | - | 6, 7 | 686 | |||
AK167528.1 | - | 6, 7 | 696 | |||
AK169224.1 | - | 6, 7 | 704 | |||
AK169244.1 | - | 6, 7 | 1204 | |||
BC012708.1 | - | 6, 7 | 702 | 4211677 | ||
BC092265.1 | - | 6, 7 | 685 | 3671250 | ||
BC094063.1 | - | 1, 2 | 59 | |||
L76223.1 | - | 6, 7 | 692 |