Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011874.2 | NP_036004.2 | 10 | 1182 | ||
GenBank mRNA | AK050677.1 | - | 10 | 1160 | ||
AK077507.1 | - | 10 | 1145 | |||
AK153991.1 | - | 10 | 1165 | |||
AK160644.1 | - | 10 | 1144 | |||
AK160718.1 | - | 10 | 1136 | |||
AK167041.1 | - | 10 | 1127 | |||
AK167528.1 | - | 10 | 1137 | |||
AK169224.1 | - | 10 | 1145 | |||
AK169244.1 | - | 10 | 1644 | |||
BC012411.1 | - | 2 | 404 | |||
BC012708.1 | - | 10 | 1143 | 4211677 | ||
BC092265.1 | - | 10 | 1126 | 3671250 | ||
BC094063.1 | - | 5 | 500 | |||
BY385516.1 | - | 2 | 174 | |||
L76223.1 | - | 10 | 1133 |