Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271544.1 | NP_001258473.1 | 19 | 2636 | ||
NM_001271545.1 | NP_001258474.1 | 18 | 2567 | |||
NM_001271546.1 | NP_001258475.1 | 19 | 2534 | |||
NM_172830.2 | NP_766418.1 | 18 | 2420 | |||
GenBank mRNA | AK050187.1 | - | 17 | 2210 | ||
AK085269.1 | - | 18 | 2420 | |||
AK085424.1 | - | 19 | 2641 | |||
AK128930.1 | - | 18 | 2776 | |||
BC113179.1 | - | 18 | 2562 | 40060674 | ||
BC113785.1 | - | 19 | 2528 |