Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173427.2 | NP_775603.2 | 1 | 2235 | ||
GenBank mRNA | AK045732.1 | - | 1 | 2238 | ||
AK050048.1 | - | 1 | 1822 | |||
AK052747.1 | - | 1 | 1805 | |||
BC043462.1 | - | 1 | 774 | |||
BC043672.1 | - | 1 | 774 |