Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Barhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005477.1 | NP_001005477.1 | 2 | 1036 | ||
GenBank mRNA | AK053799.1 | - | 2 | 1036 | ||
BC055789.1 | - | 3 | 349 | |||
BC078444.1 | - | 2 | 754 | 5688311 |