Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011776.1 | NP_001011776.1 | 1 | 96 | ||
GenBank mRNA | BC147213.1 | - | 1 | 117 | 9056261 | |
BC147214.1 | - | 1 | 117 | 9056246 |