Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146699.1 | NP_666910.1 | 1 | 578 | ||
GenBank mRNA | BC138186.1 | - | 1 | 606 | 8861205 |