Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011964.2 | NP_036094.2 | 2 | 542 | ||
GenBank mRNA | AK145915.1 | - | 1 | 1634 | ||
BC099467.1 | - | 2 | 518 | 30713077 | ||
X98111.1 | - | 2 | 542 | |||
X98112.1 | - | 2 | 542 |