Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173781.4 | NP_776142.1 | 3, 4 | 254 | ||
GenBank mRNA | AB232597.1 | - | 3, 4 | 171 | ||
AK035893.1 | - | 3, 4 | 213 | |||
AK082732.1 | - | 3, 4 | 377 | |||
BC060618.1 | - | 3, 4 | 254 | 6406134 |