Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161746.1 | NP_001155218.1 | 2 | 211 | ||
NM_013749.2 | NP_038777.2 | 3 | 316 | |||
GenBank mRNA | AF156164.1 | - | 3 | 311 | ||
AK005382.1 | - | 3 | 313 | |||
AK005530.1 | - | 3 | 312 | |||
AK160136.1 | - | 3 | 317 | |||
BC025860.1 | - | 3 | 300 | 5251646 | ||
DV655788.1 | - | 2 | 191 |