Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013749.2 | NP_038777.2 | 2 | 178 | ||
GenBank mRNA | AF156164.1 | - | 2 | 173 | ||
AK005382.1 | - | 2 | 175 | |||
AK005530.1 | - | 2 | 174 | |||
AK160136.1 | - | 2 | 179 | |||
BC025860.1 | - | 2 | 162 | 5251646 |