Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dguok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162521.1 | NP_001155993.1 | 3 | 410 | ||
NM_013764.2 | NP_038792.2 | 3 | 410 | |||
GenBank mRNA | AJ133749.1 | - | 3 | 335 | ||
AK035281.1 | - | 3 | 356 | |||
AK167071.1 | - | 2 | 171 | |||
AK185573.1 | - | 1 | 60 | |||
AK206604.1 | - | 1 | 60 | |||
AY037862.1 | - | 3 | 340 | |||
BC012647.1 | - | 3 | 355 | |||
BC094920.1 | - | 3 | 410 | 30055743 | ||
BQ746075.1 | - | 3 | 324 | |||
U90524.1 | - | 3 | 330 |