Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcl1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013775.1 | NP_038803.1 | 2 | 232 | ||
GenBank mRNA | AF195489.1 | - | 2 | 232 | ||
AK139548.1 | - | 2 | 247 | |||
BC099461.1 | - | 2 | 216 | 30935073 |