Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109988.1 | NP_001103458.1 | 4, 5 | 356 | ||
NM_001109989.2 | NP_001103459.1 | 4, 5 | 279 | |||
NM_001267717.1 | NP_001254646.1 | 4, 5 | 268 | |||
NM_053078.4 | NP_444308.1 | 3, 4 | 394 | |||
GenBank mRNA | AK020629.1 | - | 4, 5 | 217 | ||
AK048361.1 | - | 3, 4 | 194 | |||
AK081502.1 | - | 3, 4 | 195 | |||
AK133769.1 | - | 2, 3 | 1489 | |||
BC054762.1 | - | 3, 4 | 176 | 6414312 | ||
BQ715224.1 | - | 3, 4 | 181 | |||
BU057200.1 | - | 3, 4 | 184 | |||
CJ148521.1 | - | 3, 4 | 499 | |||
CJ165393.1 | - | 4, 5 | 359 | |||
CX768079.1 | - | 4, 5 | 277 | |||
EU447302.1 | - | 2, 3 | 122 | |||
X70398.1 | - | 3, 4 | 190 |