Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013879.2 | NP_038907.1 | 2, 3 | 288 | ||
GenBank mRNA | AF169152.1 | - | 2, 3 | 288 | ||
AK141231.1 | - | 2, 3 | 202 |