Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013897.2 | NP_038925.1 | 2 | 250 | ||
GenBank mRNA | AA204614.1 | - | 2 | 239 | ||
AF196314.1 | - | 239 | ||||
AK003382.1 | - | 2 | 209 | |||
AK004190.1 | - | 239 | ||||
AK012925.1 | - | 2 | 217 | |||
AK018718.1 | - | 2 | 217 | |||
AK019422.1 | - | 2 | 225 | |||
AK169258.1 | - | 2 | 229 |