Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152800.3 | NP_690013.1 | 5 | 885 | ||
GenBank mRNA | AK003976.1 | - | 1 | 73 | ||
AK162719.1 | - | 5 | 829 | |||
AK170979.1 | - | 4 | 2064 | |||
BC003466.1 | - | 5 | 870 | 2649388 | ||
BC023085.1 | - | 3 | 384 | |||
BC024469.1 | - | 5 | 848 | 4976022 |