Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256516.1 | NP_001243445.1 | 4 | 1720 | ||
NM_001256517.1 | NP_001243446.1 | 4 | 1689 | |||
NM_001256518.1 | NP_001243447.1 | 5 | 1971 | |||
NM_001256519.1 | NP_001243448.1 | 6 | 1900 | |||
NM_001256520.1 | NP_001243449.1 | 3 | 1591 | |||
NM_178761.5 | NP_848876.2 | 3 | 1559 | |||
GenBank mRNA | AK029711.1 | - | 5 | 1976 | ||
AK035113.1 | - | 3 | 1538 | |||
AK085166.1 | - | 4 | 1649 | |||
BC034706.1 | - | 4 | 1667 | 3582315 |