Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pisd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177298.3 | NP_796272.2 | 3, 4 | 313 | ||
GenBank mRNA | AK033483.1 | - | 3, 4 | 305 | ||
AK034656.1 | - | 3, 4 | 315 | |||
AK162028.1 | - | 3, 4 | 306 | |||
AK162737.1 | - | 3, 4 | 314 | |||
AK167554.1 | - | 2, 3 | 255 | |||
BC070408.1 | - | 3, 4 | 312 | 6809528 | ||
BC094594.1 | - | 3, 4 | 304 | 5707749 | ||
BC116695.1 | - | 3, 4 | 315 | 40105525 | ||
BC117509.1 | - | 3, 4 | 315 | 40105529 | ||
CX201846.1 | - | 3, 4 | 328 |