Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012402.1 | NP_001012402.1 | 2 | 811 | ||
GenBank mRNA | AK146974.1 | - | 3 | 950 | ||
AY574375.1 | - | 2 | 811 |