Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012402.1 | NP_001012402.1 | 2 | 785 | ||
GenBank mRNA | AK146974.1 | - | 3 | 924 | ||
AY574375.1 | - | 2 | 785 | |||
BC132520.1 | - | 2 | 772 | 40130797 | ||
BC138814.1 | - | 2 | 772 | 8861836 | ||
BC145424.1 | - | 2 | 742 | 9053982 | ||
BC145425.1 | - | 3 | 864 |