Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckap5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001884.1 | NP_001001884.1 | 8 | 2360 | ||
GenBank mRNA | BC072572.1 | - | 8 | 2360 | 5719876 |