Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027402.3 | NP_081678.1 | 5 | 556 | ||
GenBank mRNA | AJ401376.1 | - | 5 | 556 | ||
AK005096.1 | - | 5 | 618 | |||
BC109183.1 | - | 5 | 552 | 40050064 | ||
BC109184.1 | - | 5 | 556 | 40050065 |