Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081449.2 | NP_001074918.2 | 3 | 1279 |
Gene Symbol: Vmn2r55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104645.1 | NP_001098115.1 | 3 | 1279 | ||
GenBank mRNA | AB919108.1 | - | 3 | 1279 |
Set Membership: |
G-protein coupled receptor |