Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199016.2 | NP_950181.2 | 4 | 2973 | ||
GenBank mRNA | AK030189.1 | - | 1 | 1138 | ||
AK157622.1 | - | 4 | 2846 | |||
AK173048.1 | - | 5 | 2932 |