Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016688.2 | NP_057897.2 | 4 | 1390 | ||
GenBank mRNA | AF083929.1 | - | 4 | 1384 | ||
AK004096.1 | - | 2 | 183 | |||
AK136141.1 | - | 2 | 169 | |||
AK155050.1 | - | 4 | 1355 | |||
BC022772.1 | - | 3 | 427 | |||
DV656988.1 | - | 3 | 403 |