Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026555.2 | NP_080831.2 | 5 | 761 | ||
GenBank mRNA | AK003918.1 | - | 4 | 431 | ||
AK077943.1 | - | 5 | 701 | |||
AK082762.1 | - | 5 | 691 | |||
AK086705.1 | - | 5 | 701 | |||
AK132143.1 | - | 5 | 695 | |||
BC005487.1 | - | 4 | 598 | |||
BC025602.1 | - | 5 | 712 | 5250505 | ||
BC055903.1 | - | 5 | 708 | 4010685 | ||
BC092069.1 | - | 5 | 659 | 4215821 |