Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026819.3 | NP_081095.2 | 8 | 1002 | ||
GenBank mRNA | AK003958.1 | - | 8 | 892 | ||
AK146317.1 | - | 8 | 949 | |||
AK159964.1 | - | 8 | 924 | |||
AK168115.1 | - | 8 | 949 | |||
AK168147.1 | - | 8 | 949 | |||
BC003930.1 | - | 8 | 933 | 3485584 | ||
BC023825.1 | - | 7 | 2043 |