Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193619.1 | NP_001180548.1 | 5 | 741 | ||
NM_016887.6 | NP_058583.1 | 4 | 1063 | |||
GenBank mRNA | AF087825.1 | - | 4 | 557 | ||
AK002924.1 | - | 4 | 984 | |||
AK010558.1 | - | 4 | 983 | |||
AK087296.1 | - | 4 | 981 | |||
AK145504.1 | - | 4 | 1064 | |||
AK167250.1 | - | 4 | 984 | |||
BC008104.1 | - | 4 | 628 | 3157774 | ||
BC050007.1 | - | 4 | 976 | 6530942 | ||
BG083098.2 | - | 2 | 184 |