Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019486.1 | NP_062359.1 | 1 | 90 | ||
GenBank mRNA | BC100681.1 | - | 1 | 89 | ||
BC100683.1 | - | 1 | 88 | |||
BC100684.1 | - | 1 | 89 | |||
BC100686.1 | - | 1 | 89 | |||
BC115841.1 | - | 1 | 89 | |||
BC141882.1 | - | 1 | 90 | 40041946 | ||
BC141892.1 | - | 1 | 90 | 40041948 |