Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019580.4 | NP_062526.1 | 6 | 979 | ||
GenBank mRNA | AF212860.1 | - | 6 | 960 | ||
AK004583.1 | - | 6 | 988 | |||
AK005361.1 | - | 6 | 980 | |||
AK150807.1 | - | 6 | 1001 | |||
BC003902.1 | - | 6 | 938 | 3601420 |