Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019580.4 | NP_062526.1 | 5 | 874 | ||
GenBank mRNA | AF212860.1 | - | 5 | 855 | ||
AK004583.1 | - | 5 | 883 | |||
AK005361.1 | - | 5 | 875 | |||
AK150807.1 | - | 5 | 896 | |||
AK187418.1 | - | 2 | 168 | |||
AK198453.1 | - | 2 | 167 | |||
AK205654.1 | - | 2 | 167 | |||
BC003902.1 | - | 5 | 833 | 3601420 |