Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019953.1 | NP_064337.1 | 5 | 783 | ||
GenBank mRNA | AF186115.1 | - | 5 | 783 | ||
AK007914.1 | - | 5 | 544 | |||
AK013014.1 | - | 5 | 654 | |||
AK013568.1 | - | 5 | 525 | |||
AK019927.1 | - | 5 | 655 | |||
AK150953.1 | - | 5 | 727 | |||
BC008261.1 | - | 5 | 555 | 2650612 |