Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141931.1 | NP_001135403.1 | 6, 7 | 883 | ||
NM_001141932.1 | NP_001135404.1 | 6, 7 | 883 | |||
NM_020296.2 | NP_064692.2 | 6, 7 | 1076 | |||
GenBank mRNA | AB026569.1 | - | 6, 7 | 799 | ||
AK133054.1 | - | 6, 7 | 604 | |||
AK135280.1 | - | 6, 7 | 763 | |||
AK145897.1 | - | 6, 7 | 885 | |||
AK147109.1 | - | 6, 7 | 1077 | |||
AK161102.1 | - | 6, 7 | 833 | |||
AK161416.1 | - | 2, 3 | 213 | |||
AK164307.1 | - | 6, 7 | 877 | |||
BC016501.1 | - | 6, 7 | 1064 | 4527742 | ||
BC056609.1 | - | 3, 4 | 275 | |||
BC057866.1 | - | 6, 7 | 915 | 5327143 |